missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17257-1-AP-20UL
This item is not returnable.
View return policy
Description
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
| NDUFA3 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA3 | |
| B9, CI B9, Complex I B9, NDUFA3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4696 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95167 | |
| NDUFA3 | |
| NDUFA3 Fusion Protein Ag10912 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction