missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98152-100ul
This item is not returnable.
View return policy
Description
NDUFA3 Polyclonal antibody specifically detects NDUFA3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NDUFA3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| B9, CI-B9, complex I B9 subunit, Complex I-B9, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3 (9kD, B9), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3, 9kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3, NADH-ubiquinone oxidoreductase B9 subunit | |
| Produced in rabbits immunized with E. coli-derived Human NDUFA3 fragment. | |
| 100 μg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4696 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction