missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFA13 Monoclonal Antibody (6E1BH7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 438900
This item is not returnable.
View return policy
Description
Positive control: Human heart, Bovine heart, Rat heart, and Mouse heart isolated mitochondria, Human fibroblasts, HeLa cells.
A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.
Specifications
| NDUFA13 | |
| Monoclonal | |
| 1 mg/mL | |
| HEPES buffered saline with 0.02% sodium azide | |
| Q95KV7, Q9ERS2, Q9P0J0 | |
| Ndufa13, Ndufa13-ps1 | |
| Full-length recombinant human GRIM-19. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 6E1BH7 | |
| Unconjugated | |
| Ndufa13 | |
| 2700054G14Rik; AU022060; B16.6; CDA016; Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CGI-39; CI-B16.6; complex I B16.6 subunit; complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; gene associated with retinoic and interferon-induced mortality 19 protein; Gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM19; GRIM-19; LOC100911483; mitochondrial NADH:ubiquinone oxidoreductase B16.6 subunit; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13-like; NADH:ubiquinone oxidoreductase subunit A13; NADH-ubiquinone oxidoreductase B16.6 subunit; Ndufa13; YjeF N-terminal domain-containing protein 3; YjeF_N3; Yjefn3 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100911483, 314759, 338084, 51079, 67184 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction