missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA12 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94042-0.1ml
This item is not returnable.
View return policy
Description
NDUFA12 Polyclonal antibody specifically detects NDUFA12 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NDUFA12 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| 13 kDa differentiation-associated protein, B17.2,13kDa differentiation-associated protein, CIB17.2, CI-B17.2, complex I B17.2 subunit, Complex I-B17.2, DAP13NADH-ubiquinone oxidoreductase subunit B17.2, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12, NADH: ubiquinone oxidoreductase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human NDUFA12 (NP_061326.1). MELVQVLKRGLQQITGHGGLRGYLRVFFRTNDAKVGTLVGEDKYGNKYYEDNKQFFGRHRWVVYTTEMNGKNTFWDVDGSMVPPEWHRWLHSMTDDPPTTKPLTARKFIWTNHKFNVTGTPEQYVPYSTTRKKIQEWIPPSTPYK | |
| 0.1 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 55967 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction