missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDST3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19501
This item is not returnable.
View return policy
Description
NDST3 Polyclonal specifically detects NDST3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NDST3 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 3, EC 2.8.2.8, GlcNAc N-deacetylase/ N-sulfotransferase 3, Glucosaminyl N-deacetylase/N-sulfotransferase 3, hNDST-3, HSST3MGC130028, MGC130029, N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3, NDST-3, N-heparan sulfate sulfotransferase 3, N-HSST 3 | |
| Rabbit | |
| 101 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (92%), Rat (91%), Bovine (93%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| O95803 | |
| NDST3 | |
| Recombinant protein encompassing a sequence within the center region of human NDST3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9348 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction