missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDRG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579723
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: COLO320 whole cell. IHC: rat brain tissue, human mammary cancer tissue, human intestinal cancer tissue, mouse brain tissue.
NDRG3 (NDRG Family Member 3) is a Protein Coding gene. Diseases associated with NDRG3 includeCharcot-Marie-Tooth Disease, Type 4DandNeuropathy, Congenital Hypomyelinating Or Amyelinating, Autosomal Recessive. Among its related pathways areApoptosis and AutophagyandPI3K / Akt Signaling. An important paralog of this gene isNDRG1.
Specifications
| NDRG3 | |
| Polyclonal | |
| Unconjugated | |
| NDRG3 | |
| 4833415O14Rik; AI747193; Ndr3; NDRG family member 3; Ndrg3; N-myc downstream regulated 3; N-myc downstream regulated gene 3; N-myc downstream regulated gene 3-like; N-myc downstream-regulated gene 3 protein; Protein Ndr3; protein NDRG3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 296318, 29812, 57446 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q6AYR2, Q9QYF9, Q9UGV2 | |
| NDRG3 | |
| E.coli-derived human NDRG3 recombinant protein (Position: L181-R239). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction