missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDRG2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142707
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize reported isoform a (NP_963293.1) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
NDRG2's gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. NDRG2 is a cytoplasmic protein that may play a role in neurite outgrowth. Its gene may be involved in glioblastoma carcinogenesis.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
| NDRG2 | |
| Polyclonal | |
| Unconjugated | |
| NDRG2 | |
| AI182517; antidepressant-related protein ADRG123; AU040374; cytoplasmic protein Ndr1; DKFZp781G1938; FLJ25522; Kiaa1248; NDR1-related protein NDR2; Ndr2; NDRG family member 2; NDRG1-related protein; NDRG2; NDRG2var; N-myc downstream regulated 2; N-Myc downstream regulated gene 2; N-myc downstream regulator 2; N-myc downstream-regulated gene 2; N-myc downstream-regulated gene 2 protein; Protein Ndr2; protein NDRG2; Protein Syld709613; SYLD; syld709613 protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 171114, 29811, 57447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8VBU2, Q9QYG0, Q9UN36 | |
| NDRG2 | |
| Peptide with sequence KEAELAARILLDQGQ-C (aa 25-39). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction