missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDRG2 Antibody (4H3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43066
This item is not returnable.
View return policy
Description
NDRG2 Monoclonal antibody specifically detects NDRG2 in Human,Mouse,Porcine,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| NDRG2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 4H3 | |
| Western Blot 1:1000 - 1:10000, ELISA, Immunohistochemistry 1:500 - 1:2000 | |
| DKFZp781G1938, FLJ25522, KIAA1248cytoplasmic protein Ndr1, NDR1-related protein NDR2, NDRG family member 2, protein NDRG2, Protein Syld709613, syld709613 protein, SYLDN-myc downstream regulator 2 | |
| NDRG family member 2 - (Uniprot# Q9UN36) | |
| 100 μg | |
| Cancer | |
| 57447 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction