missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDP52 Antibody (PSH02-33), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32636
This item is not returnable.
View return policy
Description
NDP52 Monoclonal antibody specifically detects NDP52 in Human,Mouse samples. It is validated for Flow Cytometry,Western Blot
Specifications
| NDP52 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:1000 | |
| Antigen nuclear dot 52 kDa protein, calcium binding and coiled-coil domain 2, calcium-binding and coiled-coil domain-containing protein 2, NDP52MGC17318, Nuclear domain 10 protein 52, Nuclear domain 10 protein NDP52, Nuclear dot protein 52 | |
| Recombinant protein within human NDP52 aa 1-150/ 446 (Q13137). (Uniprot: Q13137) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Western Blot | |
| PSH02-33 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 10241 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction