missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDP Sensor Protein, phosphorylated

Product Code. 11513487
Change view
Click to view available options
Quantity:
10 μL
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11513487 10 μL 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11513487 Supplier Invitrogen™ Supplier No. PV3630

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

NDP Sensor Protein, phosphorylated

Recombinant D112C mutant of Myxococcus xanthus nucleoside diphosphate kinase (NDPK) labeled with the fluorophore IDCC and phosphorylated on His117
TRUSTED_SUSTAINABILITY

Specifications

Description NDP Sensor Protein, 10μL, 498μm Concentration, Universal Kinase Assay Type, Fluorescent Detection Method, Kinases Target Enzyme, –80°C Storage Temperature, Dry Ice, Used in Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation
Concentration 498μm
For Use With (Application) Protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, NDP sensor kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity
Sample Volume 10μL
Content And Storage –80°C
Product Type NDP Sensor Protein
Quantity 10 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.