missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDNL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27488-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is part of the SMC5-6 chromatin reorganizing complex and is a member of the MAGE superfamily. This is an intronless gene. Component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintece via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs).Specifications
| NDNL2 | |
| Polyclonal | |
| Unconjugated | |
| NSMCE3 | |
| HCA4, MAGE G1 antigen, MAGEG1, MAGEL3, Melanoma associated antigen G1, NDNL2, necdin like 2, Necdin like protein 2, NSE3, NSMCE3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56160 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96MG7 | |
| NSMCE3 | |
| NDNL2 Fusion Protein Ag26663 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction