missing translation for 'onlineSavingsMsg'
Learn More

NDM-1 Antibody, PerCP, Novus Biologicals™

Product Code. 18407910 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407910 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407910 Supplier Novus Biologicals Supplier No. NBP177688PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDM-1 Polyclonal antibody specifically detects NDM-1 in Mouse, Bacteria, E. coli samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDM-1
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Bacteria, E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.