missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDM-1 Antibody, PE, Novus Biologicals™
Description
NDM-1 Polyclonal antibody specifically detects NDM-1 in Mouse, Bacteria, E. coli samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NDM-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Dilution | Western Blot |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873] |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?