missing translation for 'onlineSavingsMsg'
Learn More

NDM-1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18374571 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374571 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374571 Supplier Novus Biologicals Supplier No. NBP177688AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDM-1 Polyclonal specifically detects NDM-1 in Mouse, Bacteria, E. coli samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDM-1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Mouse, Bacteria, E. coli
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.