Learn More
Invitrogen™ NDFIP2 Monoclonal Antibody (10D6D7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549299
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human HepG2 whole cell, human MCF-7 whole cell, human Hela whole cell. IHC: human placenta tissue, human squamous cell lung carcinoma tissue, human breast cancer tissue, human spleen tissue. ICC/IF: A431 cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Activates HECT domain-containing E3 ubiquitin-protein ligases, including ITCH, NEDD4, NEDD4L, SMURF2, WWP1 and WWP2, and consequently modulates the stability of their targets. As a result, may control many cellular processes. Recruits ITCH, NEDD4 and SMURF2 to endosomal membranes. Negatively regulates KCNH2 potassium channel activity by decreasing its cell-surface expression and interfering with channel maturation through recruitment of NEDD4L to the Golgi apparatus and multivesicular body where it mediates KCNH2 degradation (PubMed:26363003). May modulate EGFR signaling. Together with NDFIP1, limits the cytokine signaling and expansion of effector Th2 T-cells by promoting degradation of JAK1, probably by ITCH- and NEDD4L-mediated ubiquitination. [UniProt].
Specifications
| NDFIP2 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q9NV92 | |
| NDFIP2 | |
| E.coli-derived human NDFIP2 recombinant protein (Position: M16-L336). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 10D6D7 | |
| Unconjugated | |
| NDFIP2 | |
| 0710001O20Rik; 2810436B12Rik; 9130207N19Rik; KIAA1165; MAPK-activating protein PM04 PM05 PM06 PM07; mKIAA1165; N4wbp5a; NDFIP2; Nedd4 family interacting protein 2; NEDD4 family-interacting protein 2; NEDD4 WW domain-binding protein 5A; NF-kappa-B-activating protein 413; putative MAPK-activating protein PM04/PM05/PM06/PM07; Putative NF-kappa-B-activating protein 413; Unknown (protein for IMAGE:7958216); wu:fa92h03; wu:fk31f07; zgc:63948 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 54602 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.