missing translation for 'onlineSavingsMsg'
Learn More

NDFIP1, Mouse anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16160337
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16160337 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16160337 Supplier Abnova Supplier No. H00080762B01P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Sequence: MALALAALAAVEPACGSRYQQLQNEEESGEPEQAAGDAPPPYSSISAESAAYFDYKDESGFPKPPSYNVATTLPSYDEAERTKAEATIPLVPGRDEDFVGRDDFDDADQLRIGNDGIFMLTFFMAFLFNWIGFFLSFCLTTSAAGRYGAISGFGLSLIKWILIVRFSTYFPGYFDGQYWLWWVFLVLGFLLFLRGFINYAKVRKMPETFSNLPRTRVLFIY

Specifications

Antigen NDFIP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Nedd4 family interacting protein 1
Gene Accession No. NM_030571.2
Gene Alias MGC10924/N4WBP5
Gene Symbols NDFIP1
Host Species Mouse
Immunogen NDFIP1 (NP_085048.1, 1 a.a. ∼ 221 a.a) full-length human protein.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80762
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.