missing translation for 'onlineSavingsMsg'
Learn More

NDFIP1 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30736342 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736342 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736342 Supplier Novus Biologicals Supplier No. AF8246Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

NDFIP1 Polyclonal antibody specifically detects NDFIP1 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDFIP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Breast cancer-associated protein SGA-1M, MGC10924, N4WBP5Putative NF-kappa-B-activating protein 164, Nedd4 family interacting protein 1, NEDD4 family-interacting protein 1, NEDD4 WW domain-binding protein 5, Putative MAPK-activating protein PM13, Putative NFKB and MAPK-activating protein
Host Species Sheep
Immunogen E. coli-derived recombinant human NDFIP1, Ala10-Asp105, Accession # Q9BT67
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80762
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.