missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Boc-Nalpha-Fmoc-L-ornithine, 96%, Thermo Scientific Chemicals

Product Code. 15489278
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109425-55-0
Molecular Formula C25H30N2O6
Molecular Weight (g/mol) 454.523
MDL Number MFCD00065668
InChI Key JOOIZTMAHNLNHE-NRFANRHFSA-N
Synonym fmoc-orn boc-oh, nalpha-fmoc-ndelta-boc-l-ornithine, fmoc-l-orn boc-oh, ndelta-boc-nalpha-fmoc-l-ornithine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2s-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-orn boc, pubchem10032, n-a-boc-n-deta-boc-l-ornithine, n alpha-fmoc-n delta-boc-l-ornithine
PubChem CID 2756114
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 111°C to 115°C
Beilstein 4772025
Formula Weight 454.52
Percent Purity 96%
Quantity 1 g
Chemical Name or Material Ndelta-Boc-Nalpha-Fmoc-L-ornithine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'