missing translation for 'onlineSavingsMsg'
Learn More

Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine, 98%, Thermo Scientific™

Product Code. 15498808
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498808 1 g 1g
15408818 5 g 5g
15418818 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498808 Supplier Thermo Scientific Alfa Aesar Supplier No. H62268.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2480-93-5
Molecular Formula C18H26N2O6
Molecular Weight (g/mol) 366.414
MDL Number MFCD00038259
InChI Key QYYCZJUFHDLLOJ-AWEZNQCLSA-N
Synonym boc-orn z-oh, s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-orn cbz-oh, nalpha-boc-ndelta-cbz-l-ornithine, nalpha-tert-butoxycarbonyl-ndelta-carbobenzoxy-l-ornithine, 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-phenylmethoxycarbonylamino pentanoic acid, 2s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-tert-butoxycarbonyl-n'-benzyloxycarbonyl-l-ornithine, n-alpha-boc-n-delta-z-l-ornithine, bocnh-orn z-oh
PubChem CID 7269338
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Beilstein 2783761
Quantity 1 g
Formula Weight 366.41
Percent Purity 98%
Chemical Name or Material Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.