missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554972
This item is not returnable.
View return policy
Description
Immunogen sequence: KDEARDLRQE LAVQQKQEKP RTPMPSSVEA ERTDTAVQAT GSVPSTPIAH RGPSSSLNTP GSFRRGLDDS TGGTPLTPAA RISALNIVGD LLRKVGALES KLAS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 83%.
NDE1 is s a member of the nuclear distribution E (NudE) family of proteins. It is localized at the centrosome and interacts with other centrosome components as part of a multiprotein complex that regulates dynein function. NDE1 protein plays an essential role in microtubule organization, mitosis and neuronal migration. Mutations to the gene lead to Lissencephaly 4 (LIS4) and Microhydranencephaly (MHAC).
Specifications
| NDE1 | |
| Polyclonal | |
| Unconjugated | |
| NDE1 | |
| 2810027M15Rik; AU042936; AW822251; HOM-TES-87; LIS1-interacting protein NUDE1; LIS1-interacting protein NUDE1, rat homolog; LIS4; MHAC; mNudE; NDE; NDE1; nuclear distribution gene E homolog 1; nuclear distribution gene E homolog 1 (A nidulans); nuclear distribution protein nudE homolog 1; NudE; nudE neurodevelopment protein 1; nudE nuclear distribution E homolog 1; nudE nuclear distribution gene E homolog 1; Nude1; rNudE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54820, 67203, 83836 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9CZA6, Q9ES39, Q9NXR1 | |
| NDE1 | |
| Recombinant protein corresponding to Human NDE1. Recombinant protein control fragment (Product #RP-92277). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction