missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDE1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-06537V
This item is not returnable.
View return policy
Description
NDE1 Polyclonal antibody specifically detects NDE1 in Human samples. It is validated for Western Blot
Specifications
| NDE1 | |
| Polyclonal | |
| Western Blot | |
| FLJ20101, HOM-TES-87, nuclear distribution protein nudE homolog 1, NudE, nudE nuclear distribution gene E homolog 1 (A. nidulans), NUDE1, rat homolog | |
| The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human nuclear distribution gene E using the numbering given in entry NP_060138.1 (GeneID 54820). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 54820 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction