missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDC80 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38149-20ul
This item is not returnable.
View return policy
Description
NDC80 Polyclonal antibody specifically detects NDC80 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| NDC80 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| HEC1KNTC2, HECRetinoblastoma-associated protein HEC, Highly expressed in cancer protein, highly expressed in cancer, rich in leucine heptad repeats, highly expressed in cancer, rich in leucine heptad repeats (yeast), hsHec1, hsNDC80, kinetochore associated 2, Kinetochore protein Hec1, kinetochore protein NDC80 homolog, Kinetochore-associated protein 2, NDC80 homolog, kinetochore complex component (S. cerevisiae), TID3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 343-642 of human NDC80 (NP_006092.1).,, Sequence:, ETIKQENTRLQNIIDNQKYSVADIERINHERNELQQTINKLTKDLEAEQQKLWNEELKYARGKEAIETQLAEYHKLARKLKLIPKGAENSKGYDFEIKFNPEAGANCLVKYRAQVYVPLKELLNETEEEINKALNKKMGLEDTLEQLNAMITESKRSVRTLKEEVQKLDDLYQQKIKEAEEEDEKCASELESLEKHKHLLESTVNQGLSEAMNELDAVQREYQLVVQTTTEERRKVGNNLQRLLEMVATHVGSVEKHLEEQIAKVDREYEECMSEDLSENIKEIRDKYEKKATLIKSSEE | |
| 20 μL | |
| Cell Biology, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Mitotic Regulators, modENCODE Antibodies, Stem Cell Markers | |
| 10403 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction