missing translation for 'onlineSavingsMsg'
Learn More

NDC80 Antibody (1A10), Novus Biologicals™

Product Code. 18393019 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393019 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393019 Supplier Novus Biologicals Supplier No. H00010403M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDC80 Monoclonal antibody specifically detects NDC80 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDC80
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 1A10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH35617
Gene Alias HEC1KNTC2, HECRetinoblastoma-associated protein HEC, Highly expressed in cancer protein, highly expressed in cancer, rich in leucine heptad repeats, highly expressed in cancer, rich in leucine heptad repeats (yeast), hsHec1, hsNDC80, kinetochore associated 2, Kinetochore protein Hec1, kinetochore protein NDC80 homolog, Kinetochore-associated protein 2, NDC80 homolog, kinetochore complex component (S. cerevisiae), TID3
Host Species Mouse
Immunogen KNTC2 (AAH35617, 545 a.a. ~ 642 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TVNQGLSEAMNELDAVQREYQLVVQTTTEERRKVGNNLQRLLEMVATHVGSVEKHLEEQIAKVDREYEECMSEDLSENIKEIRDKYEKKATLIKSSEE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10403
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.