missing translation for 'onlineSavingsMsg'
Learn More

NCX 1000, MedChemExpress

Product Code. 16453099
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453099 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16453099 Supplier Medchem Express Supplier No. HYU000231mg
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NCX 1000 is a liver-specific NO donor compound derived from ursodeoxycholic acid (UDCA).

NCX 1000 is a liver-specific NO donor compound derived from ursodeoxycholic acid (UDCA).

Specifications

Chemical Name or Material NCX 1000
CAS 401519-96-8
Color White
Purity Grade Notes Research
Molecular Formula C38H55NO10
Quantity 1 mg
SMILES C[C@H](CCC(OC1=CC=C(/C=C/C(OCCCCO[N+]([O-])=O)=O)C=C1OC)=O)[C@H]2CC[C@@]3([H])[C@]4([H])[C@@H](O)C[C@]5([H])C[C@H](O)CC[C@]5(C)[C@@]4([H])CC[C@]23C
Molecular Weight (g/mol) 685.84
Formula Weight 685.84
Percent Purity 98.06%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.