missing translation for 'onlineSavingsMsg'
Learn More

NCT-502, MedChemExpress

Product Code. 16487617
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487617 5 mg 5mg
16474868 10 mg 10mg
16473266 25 mg 25mg
16415656 100 mg 100mg
16411127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487617 Supplier Medchem Express Supplier No. HY117240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NCT-502 is a human phosphoglycerate dehydrogenase (PHGDH) inhibitor, cytotoxic to PHGDH-dependent cancer cells, and reduces glucose-derived serine production, with an IC50 of 3.7 μM against PHGDH.

NCT-502 is a human phosphoglycerate dehydrogenase (PHGDH) inhibitor, cytotoxic to PHGDH-dependent cancer cells, and reduces glucose-derived serine production, with an IC50 of 3.7 μM against PHGDH.

Specifications

Chemical Name or Material NCT-502
CAS 1542213-00-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H20F3N5S
Quantity 5 mg
Solubility Information DMSO : 130 mg/mL (328.74 mM; Need ultrasonic)
SMILES S=C(NC1=NC(C)=CC(C)=C1)N2CCN(CC2)C3=NC=C(C=C3)C(F)(F)F
Molecular Weight (g/mol) 395.45
Formula Weight 395.45
Percent Purity 99.18%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.