missing translation for 'onlineSavingsMsg'
Learn More

NCT-501, MedChemExpress

Product Code. 16473637
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473637 50 mg 50mg
16457536 100 mg 100mg
16417628 10 mg 10mg
16410098 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473637 Supplier Medchem Express Supplier No. HY18768

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NCT-501 is a potent and selective theophylline-based inhibitor of aldehyde dehydrogenase 1A1 (ALDH1A1), inhibits hALDH1A1 with IC50 of 40 nM, typically shows better selectivity over other ALDH isozymes and other dehydrogenases (hALDH1B1, hALDH3A1, and hALDH2, IC50 >57 μM).

NCT-501 is a potent and selective theophylline-based inhibitor of aldehyde dehydrogenase 1A1 (ALDH1A1), inhibits hALDH1A1 with IC50 of 40 nM, typically shows better selectivity over other ALDH isozymes and other dehydrogenases (hALDH1B1, hALDH3A1, and hALDH2, IC50 >57 μM).

Specifications

Chemical Name or Material NCT-501
CAS 1802088-50-1
Color White
Purity Grade Notes Research
Molecular Formula C21H32N6O3
Quantity 50 mg
Solubility Information DMSO : ≥ 28 mg/mL (67.22 mM)
SMILES O=C(N1C)N(C)C2=C(N(CCC(C)C)C(CN3CCN(C(C4CC4)=O)CC3)=N2)C1=O
Molecular Weight (g/mol) 416.52
Formula Weight 416.52
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.