missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCRNA00114 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569384
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
LINC00114 is a protein coding gene.
Specifications
| NCRNA00114 | |
| Polyclonal | |
| Unconjugated | |
| LINC00114 | |
| C21orf24; LINC00114; long intergenic non-protein coding RNA 114; NCRNA00114 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 400866 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6XXX2 | |
| LINC00114 | |
| synthetic peptide directed towards the N terminal region of human NCRNA00114. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction