missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Porablot Nitrocellulose membrane for blotting applications

Product Code. 12807411
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12807411 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12807411 Supplier Macherey-Nagel™ Supplier No. 741280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nitrocellulose membrane for high quality transfer membranes for biomolecule analysis.

For membrane selection and specifications, see 'Summary of porablot membrane applicability' table.

porablot NCP

The classical transfer medium for binding proteins and nucleic acids. Membrane material: 100% nitrocelluloseMembrane character: HydrophilicPore size: 0.45μmBinding capacity: 100μg/cm2Applications: Southern, Northern and Western blotting, colony and plaque transfer, dot blottingAlso available with supporting tissue, see porablot NCL.

porablot NCL

The classical transfer medium for binding proteins and nucleic acids - the supporting tissue prevents breaking, tearing, or shrinking.

  • Membrane material: 100% nitrocellulose with inert supporting tissue
  • Membrane character: Hydrophilic
  • Pore size: 0.45μm
  • Binding capacity: 100μg/cm2
  • Applications: Southern, Northern and Western blotting, colony and plaque transfer, dot blottingAlso available without supporting tissue, see porablot NCP.

porablot NY amp

The amphoteric nylon membrane with supporting tissue for improved handling.

  • Membrane material: High purity nylon with inert supporting tissue
  • Membrane character: Amphoteric (isoelectric point at pH6.5)
  • Pore size: 0.20μm
  • Applications: Southern and Northern blotting, colony and plaque lifts, dot blottingAlso available with positive surface charge, see porablot NY plus.

porablot NY plus

The strongly cationic nylon membrane with supporting tissue for improved handling.

  • Membrane material: High purity nylon with inert supporting tissue
  • Membrane character: Cationic (in the pH range of 3 to 10)
  • Pore size: 0.45μm
  • Applications: Southern, Northern and Western blotting. Very well suited for electro- and vacuum blotting techniquesAlso available with positive surface charge, see porablot NY amp.

porablot PVDF

The classical transfer membrane for Western transfer, protein binding assays.

  • Membrane material: Polyvinylidene difluoride (PVDF)
  • Membrane character: Hydrophobic
  • Pore size: 0.20μm
  • Binding capacity: 50μg/cm2 to 100μg/cm2 (For large, globular proteins such as immunoglobulins, for smaller peptides the binding capacity is correspondingly larger)
  • Applications: Western blotting, protein binding assays, protein sequencing
  • Transfer membranes from a variety of polymeric materials with a broad range of pore sizes and surface chemistry
  • Easy analysis and characterisation of biomolecules
  • Conservation of expensive test solutions
  • Possibility of multiple investigations with one blot
  • Concentration of a sample with consequent higher detection sensitivity
  • Convenient storage of results
  • Outstanding band resolution due to uniform, carefully controlled pore structure
  • Available in custom sizes – contact our Technical Support Team for further information Transfer membranes for DNA, RNA, bacterial colonies and protein blotting applications.

Specifications

For Use With (Application) Southern, Northern, and Western blotting, colony and plaque transfer, dot blotting
Binding Capacity 100 μg/cm2
Code 86
Quantity 1 Ea.
Material NCP
Pore Size 0.45 μm
Content And Storage 1 Ea., 15°C to 25°C
compliance-icons
Product Identifier
  • Porablot NCP (0.3x3 m, 1 roll)
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.