missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-5802
This item is not returnable.
View return policy
Description
NCOR2 Polyclonal antibody specifically detects NCOR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
Specifications
| NCOR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/mL BSA | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 4 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| CTG repeat protein 26, CTG26nuclear receptor co-repressor 2, FLJ25011, N-CoR2, nuclear receptor corepressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, SMAP270, SMRTE, SMRTE-tau, SMRTT3 receptor-associating factor, TNRC14, TRAC, TRAC1 | |
| His-tagged fusion protein corresponding to amino acid residues 95-565 from human SMRT. | |
| 0.1 mg | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 9612 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction