missing translation for 'onlineSavingsMsg'
Learn More

NCOR2 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30737651 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737651 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737651 Supplier Novus Biologicals Supplier No. AF7017E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

NCOR2 Polyclonal antibody specifically detects NCOR2 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCOR2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CTG repeat protein 26, CTG26nuclear receptor co-repressor 2, FLJ25011, N-CoR2, nuclear receptor corepressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, SMAP270, SMRTE, SMRTE-tau, SMRTT3 receptor-associating factor, TNRC14, TRAC, TRAC1
Host Species Sheep
Immunogen E. coli-derived recombinant human NCOR2, Lys2268-Arg2449, Accession # Q9Y618
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9612
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.