missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCOA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101726
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NCOA7.
NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
Specifications
| NCOA7 | |
| Polyclonal | |
| Unconjugated | |
| NCOA7 | |
| 140 kDa estrogen receptor-associated protein; 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nbla00052; Nbla10993; Ncoa7; NCOA7-AS; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; RGD1566426; TBC/LysM-associated domain containing 4; TLDC4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 135112 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NI08 | |
| NCOA7 | |
| A synthesized peptide derived from human NCOA7(Accession Q8NI08), corresponding to amino acid residues E421-G471. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction