missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Concentration | 0.2 mg/ml |
| For Use With (Application) | Antibody Competition |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin |
| Gene ID (Entrez) | 8031 |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator |
| Gene Symbol | NCOA4 |
| Protein | NCOA4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?