missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA3/AIB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77785
This item is not returnable.
View return policy
Description
NCOA3/AIB1 Polyclonal specifically detects NCOA3/AIB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.
Specifications
| NCOA3/AIB1 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| ACTRCBP-interacting protein, AIB1AIB-1, Amplified in breast cancer 1 protein, BHLHE42, bHLHe42p/CIP, CAGH16, Class E basic helix-loop-helix protein 42, CTG26, KAT13B, MGC141848, NCoA-3, nuclear receptor coactivator 3, pCIP RAC-3, RAC3pCIP, Receptor-associated coactivator 3, SRC3, SRC-3EC 2.3.1.48, Steroid receptor coactivator protein 3, Thyroid hormone receptor activator molecule 1, TNRC16, TRAM1, TRAM-1TNRC14 | |
| Rabbit | |
| 155.29 kDa | |
| 0.1 mg | |
| Breast Cancer, Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 8202 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 1.24 mg/ml | |
| Western Blot 1:500-1:1000, ELISA 1:175000, Immunohistochemistry 5ug/ml, Immunoprecipitation 1:100 | |
| Q9Y6Q9 | |
| NCOA3 | |
| This affinity purified NCOA3/AIB1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human NCOA3/AIB1/AIB1 isoform a protein. (Uniprot: Q9Y6Q9) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody reacts with several isoforms of human NCOA3/AIB1 protein (a, b, d, e and f). | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction