missing translation for 'onlineSavingsMsg'
Learn More

NCOA2 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30737685 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737685 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737685 Supplier Novus Biologicals Supplier No. AF8010L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

NCOA2 Polyclonal antibody specifically detects NCOA2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCOA2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias BHLHE75, bHLHe75MGC138808, Class E basic helix-loop-helix protein 75, EC 2.3.1.48, GRIP1glucocorticoid receptor-interacting protein-1, KAT13C, NCoA-2SRC2, nuclear receptor coactivator 2, TIF2hTIF2, Transcriptional intermediary factor 2
Host Species Sheep
Immunogen E. coli-derived recombinant human NCOA2, Phe1090-Leu1247, Accession # Q15596
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10499
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.