missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCKIPSD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00167-100ul
This item is not returnable.
View return policy
Description
NCKIPSD Polyclonal antibody specifically detects NCKIPSD in Human samples. It is validated for ELISA
Specifications
| NCKIPSD | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| 54 kDa vimentin-interacting protein, 90 kDa SH3 protein interacting with Nck, AF3p21, AF3P21DIP-1, dia interacting protein, Dia-interacting protein 1, diaphanous protein interacting protein, Diaphanous protein-interacting protein, DIP, DIP1, MGC23891, NCK interacting protein with SH3 domain, ORF1, SPIN90VIP54, WASLBP, WASP-interacting SH3-domain protein, WISHSH3 adapter protein SPIN90, Wisk90 kDa | |
| Produced in rabbits immunized with purified, recombinant Human NCKIPSD (Uniprot#: Q9NZQ3-3; Met1-Thr244) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 51517 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction