missing translation for 'onlineSavingsMsg'
Learn More

NCKIPSD Antibody (1A10), Novus Biologicals™

Product Code. 18359749 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359749 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359749 Supplier Novus Biologicals Supplier No. H00051517M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NCKIPSD Monoclonal antibody specifically detects NCKIPSD in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCKIPSD
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_909119
Gene Alias 54 kDa vimentin-interacting protein, 90 kDa SH3 protein interacting with Nck, AF3p21, AF3P21DIP-1, dia interacting protein, Dia-interacting protein 1, diaphanous protein interacting protein, Diaphanous protein-interacting protein, DIP, DIP1, MGC23891, NCK interacting protein with SH3 domain, ORF1, SPIN90VIP54, WASLBP, WASP-interacting SH3-domain protein, WISHSH3 adapter protein SPIN90, Wisk90 kDa
Host Species Mouse
Immunogen NCKIPSD (NP_909119.1, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MYRALYAFRSAEPNALAFAAGETFLVLERSSAHWWLAARARSGETGYVPPAYLRRLQGLEQDVLQAIDRAIEAVHNTAMRDGGKYSLEQRGVLQKLIHH
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51517
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.