missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCKAP5L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559404
This item is not returnable.
View return policy
Description
Immunogen sequence: EEKVMKGIEE NVLRLQGQER APGAEVKHRN TSSIASWFGL KKSKLPALNR RTEATKNKEG AGGGSPLRRE VKMEARKL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
KIAA1602, also named as NCKAP5L, has 4 isoforms. The target band is about 139-145 kDa for phosphorylation.
Specifications
| NCKAP5L | |
| Polyclonal | |
| Unconjugated | |
| NCKAP5L | |
| C230021P08Rik; Centrosomal protein of 169 kDa; Cep169; FP1193; KIAA1602; NCK associated protein 5 like; NCKAP5L; NCKAP5-like; NCK-associated protein 5-like; RGD1561571 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57701 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HCH0 | |
| NCKAP5L | |
| Recombinant protein corresponding to Human NCKAP5L. Recombinant protein control fragment (Product #RP-97809). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction