missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCK2 Recombinant Rabbit Monoclonal Antibody (24GB105)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557099
This item is not returnable.
View return policy
Descripción
NCK2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The NCK family of SH2/SH3 adaptor proteins consists of two members, NCK1 and NCK2, which couple tyrosine kinase signaling, including the EGF and PDGF receptor-pathways, to downstream signaling proteins. The NCK2 protein binds human PDGFR-b (Tyr-1009); overexpression of NCK2 inhibits PDGF-induced membrane ruffling and lamellipod formation.
Especificaciones
| NCK2 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43639 | |
| NCK2 | |
| A synthesized peptide derived from human Nck2 (aa 50-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB105 | |
| Unconjugated | |
| NCK2 | |
| 4833426I10Rik; cytoplasmic protein NCK2; Grb4; growth factor receptor-bound protein 4; NCK adaptor protein 2; NCK2; Nck-2; NCKbeta; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8440 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido