missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCK2 Rabbit anti-Human, Mouse, Rat, Clone: S01-3C6, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | NCK2 |
|---|---|
| Clone | S01-3C6 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313314
|
Novus Biologicals
NBP3-15063-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18377471
|
Novus Biologicals
NBP3-15063-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NCK2 Monoclonal antibody specifically detects NCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NCK2 | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| GRB4, NCK adaptor protein 2cytoplasmic protein NCK2, Nck-2, NCKbeta, noncatalytic region of tyrosine kinase, beta, SH2/SH3 adaptor protein NCK-beta | |
| A synthetic peptide of human NCK2 (Uniprot#: O43639) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-3C6 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 8440 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title