missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104540
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NCK2.
The NCK family of SH2/SH3 adaptor proteins consists of two members, NCK1 and NCK2, which couple tyrosine kinase signaling, including the EGF and PDGF receptor-pathways, to downstream signaling proteins. The NCK2 protein binds human PDGFR-b (Tyr-1009); overexpression of NCK2 inhibits PDGF-induced membrane ruffling and lamellipod formation.
Specifications
| NCK2 | |
| Polyclonal | |
| Unconjugated | |
| NCK2 | |
| 4833426I10Rik; cytoplasmic protein NCK2; Grb4; growth factor receptor-bound protein 4; NCK adaptor protein 2; NCK2; Nck-2; NCKbeta; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17974, 8440 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43639, O55033 | |
| NCK2 | |
| A synthesized peptide derived from human NCK2(Accession O43639), corresponding to amino acid residues Y164-L214. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction