missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15247-1-AP-20UL
This item is not returnable.
View return policy
Description
NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.Specifications
| NCK1 | |
| Polyclonal | |
| Unconjugated | |
| NCK1 | |
| Cytoplasmic protein NCK1, NCK, Nck 1, NCK adaptor protein 1, NCK1, NCKalpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17973, 300955, 4690 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16333, Q99M51 | |
| Nck1 | |
| NCK1 Fusion Protein Ag0979 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction