Learn More
Invitrogen™ NCK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583157
Description
Immunogen sequence: MAEEVVVVAK FDYVAQQEQE LDIKKNERLW LLDDSKSWWR VRNSMNKTGF VPSNYVERKN SARKASIVKN LKDTLGIGKV KRKPSVPDSA SPADDSFVDP GERLYDLNMP AYVKFNYMAE REDELSLIKG TKVIVMEKCS DGWWRGSYNG Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 92%.
NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
| NCK1 | |
| Polyclonal | |
| Unconjugated | |
| Nck1 | |
| 6330586M15Rik; Cytoplasmic protein NCK1; D230010O13Rik; melanoma NCK protein; MGC12668; Nck; NCK adaptor protein 1; NCK tyrosine kinase; NCK1; nck-1; NCKalpha; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P16333 | |
| Nck1 | |
| Recombinant protein corresponding to Human NCK1. Recombinant protein control fragment (Product #RP-104619). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.