missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCK1 Recombinant Rabbit Monoclonal Antibody (4R1N9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535758
This item is not returnable.
View return policy
Description
Immunogen sequence: PFSSSNDEEL NFEKGDVMDV IEKPENDPEW WKCRKINGMV GLVPKNYVTV MQNNPLTSGL EPSPPQCDYI RPSLTGKFAG NPWYYGKVTR HQAEMALNER G.
NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
| NCK1 | |
| Recombinant Monoclonal | |
| 0.38 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16333, Q99M51 | |
| Nck1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human NCK11 (P16333). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4R1N9 | |
| Unconjugated | |
| Nck1 | |
| 6330586M15Rik; Cytoplasmic protein NCK1; D230010O13Rik; melanoma NCK protein; MGC12668; Nck; NCK adaptor protein 1; NCK tyrosine kinase; NCK1; nck-1; NCKalpha; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17973, 300955, 4690 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction