missing translation for 'onlineSavingsMsg'
Learn More

NCGC00247743, MedChemExpress

Product Code. 16437337
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16437337

Brand: Medchem Express HY112308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NCGC00247743 is a histone lysine demethylase KDM4 inhibitor.

NCGC00247743 is a histone lysine demethylase KDM4 inhibitor.

Specifications

Chemical Name or Material NCGC00247743
CAS 1435192-04-3
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C24H29N3O2
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (159.64 mM; Need ultrasonic)
SMILES OC1=C(N=CC=C2)C2=CC(C3=CC(C(NCCCCCCN(C)C)=O)=CC=C3)=C1
Molecular Weight (g/mol) 391.51
Formula Weight 391.51
Percent Purity 96.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.