missing translation for 'onlineSavingsMsg'
Learn More

NCF4 Antibody (OTI4B5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-72893F

Product Code. 18163896

  • £595.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NCF4 Monoclonal specifically detects NCF4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCF4
Monoclonal
FITC
PBS with 0.05% Sodium Azide
NCF4
Full length recombinant protein of human NCF4 (NP_000622) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Immunohistochemistry
OTI4B5
Immunohistochemistry
MGC3810, NCF, NCF-4, neutrophil cytosol factor 4, neutrophil cytosolic factor 4 (40kD), neutrophil cytosolic factor 4, 40kDa, Neutrophil NADPH oxidase factor 4, p40phox, p40-phox, SH3 and PX domain-containing protein 4, SH3PXD4P40PHOX
Mouse
Affinity Purified
Signal Transduction
4689.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.