missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCF2 293T Cell Overexpression Lysate (Denatured), Abnova
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | NCF2 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC001606 |
| Molecular Weight (g/mol) | 57.97 |
| pH Range | 6.8 |
| Plasmid | pCMV-NCF2 full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-NCF2 antibody (H00004688-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Safety and Handling
Product Identifier
- 100UL NCF2 293T Cell Transient Overexpression Lysa
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Mercaptoethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction