missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCEH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550285
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total NCEH1 protein.
Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.
Specifications
| NCEH1 | |
| Polyclonal | |
| Unconjugated | |
| NCEH1 | |
| AADACL1; Arylacetamide deacetylase-like 1; B230106I24Rik; chlorpyrifos oxon-binding protein; CPO-BP; Kiaa1363; NCEH; NCEH1; Neutral cholesterol ester hydrolase 1; RGD1311104 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 320024, 57552 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6PIU2, Q8BLF1 | |
| NCEH1 | |
| Fusion protein corresponding to residues near the C terminal of human Neutral cholesterol ester hydrolase 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido