missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCEH1 Polyclonal Antibody

Beskrivning
Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.
Specifikationer
Specifikationer
| Antigen | NCEH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | NCEH1 |
| Gene Accession No. | Q6PIU2 |
| Gene Alias | AADACL1; Arylacetamide deacetylase-like 1; B230106I24Rik; chlorpyrifos oxon-binding protein; CPO-BP; Kiaa1363; NCEH; NCEH1; Neutral cholesterol ester hydrolase 1; RGD1311104 |
| Gene Symbols | NCEH1 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?