missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCCRP1 Antibody (5C.6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2071
This item is not returnable.
View return policy
Description
NCCRP1 Monoclonal specifically detects NCCRP1 in Rat, Porcine, Bovine, Canine, Equine, Fish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Inhibition Activity.
Specifications
| NCCRP1 | |
| Monoclonal | |
| 0.73 mg/mL | |
| Western Blot 10 ug/ml, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Inhibition Activity | |
| LOC342897, NCCRP-1, nonspecific cytotoxic cell receptor protein 1 homolog, non-specific cytotoxic cell receptor protein 1 homolog, non-specific cytotoxic cell receptor protein 1 homolog (zebrafish) | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Nonspecific Cytotoxic Cell (5C.6) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Inhibition Assays | |
| 5C.6 | |
| Unconjugated | |
| Q6ZVX7 | |
| NCCRP1 | |
| Purified NCC cells from catfish. | |
| 200 μg | |
| Adaptive Immunity, Immunology | |
| 342897 | |
| Rat, Pig, Bovine, Canine, Equine, Fish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction