missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCBP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA597995

Product Code. 16540822

  • £424.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NCBP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5' cap of nascent pre-mRNA in the nucleoplasm. The encoded protein has an RNP domain commonly found in RNA binding proteins, and contains the cap-binding activity. The CBC promotes pre-mRNA splicing, 3'-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCBP2
Polyclonal
Unconjugated
NCBP2
20 kDa nuclear cap-binding protein; 20kDa; 5930413E18Rik; AA536802; AI843301; C79367; CBC2; CBP20; cell proliferation-inducing gene 55 protein; NCBP 20 kDa subunit; NCBP interacting protein 1; NCBP2; NCBP-interacting protein 1; NIP1; nuclear cap binding protein subunit 2; nuclear cap binding protein subunit 2, 20kD; nuclear cap binding protein subunit 2, 20kDa; nuclear cap-binding protein subunit 2; phosphatidylinositol glycan anchor biosynthesis, class Z; PIG55; Pigz
Rabbit
Antigen Affinity Chromatography
RUO
22916, 68092
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.71 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52298, Q9CQ49
NCBP2
Recombinant Human Nuclear cap-binding protein subunit 2 protein (1-156AA).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.